2,6-Difluoro-3-methylbenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1548143-20-9
:
Formula:C₇H₅ClF₂O₂S
MDL Number:MFCD24310152
:226.628 g/mol
Storage Temperature:Ambient
UN:1760
ADR:8,III
Frequently Bought Together