2,3-Difluoro-4-methoxybenzenesulphonyl chloride
Catalog # APOSPC303198-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:242.628 g/mol
Storage Temperature:Ambient
CAS Number:1341660-99-8
Formula:C₇H₅ClF₂O₃S
MDL Number:MFCD17282234