
3,4-Dichloro-5-(trifluoromethyl)benzyl bromide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1823323-67-6
:
Formula:C₈H₄BrCl₂F₃
MDL Number:MFCD28103368
:307.923 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together