Order Entry
Czech Republic
ContactUsLinkComponent
2,6-Dichloro-3-(trifluoromethyl)benzenesulphonyl chloride
2,6-Dichloro-3-(trifluoromethyl)benzenesulphonyl chloride
Catalog # APOSPC303178-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dichloro-3-(trifluoromethyl)benzenesulphonyl chloride
Catalog # APOSPC303178-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :313.509 g/mol
Storage Temperature:Ambient
CAS Number:1706458-88-9
Formula:C₇H₂Cl₃F₃O₂S
MDL Number:MFCD28054229