3,4-Dichloro-5-(trifluoromethoxy)acetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1823541-26-9
:
Formula:C₉H₅Cl₂F₃O₂
MDL Number:MFCD28103364
:273.036 g/mol
Storage Temperature:Ambient
Frequently Bought Together