3-Butoxy-2,6-difluorobenzyl bromide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1706446-25-4
:
Formula:C₁₁H₁₃BrF₂O
MDL Number:MFCD27986842
:279.121 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
Frequently Bought Together