
2,5-Dichloro-3-(trifluoromethyl)aniline 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:103995-81-9
:
Formula:C₇H₄Cl₂F₃N
MDL Number:MFCD27986858
:230.015 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together