Order Entry
Czech Republic
ContactUsLinkComponent
3-(2,3,4-Trifluorophenyl)propionic acid 97%
3-(2,3,4-Trifluorophenyl)propionic acid 97%
Catalog # APOSPC302935-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(2,3,4-Trifluorophenyl)propionic acid 97%
Catalog # APOSPC302935-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :204.146 g/mol
Storage Temperature:Ambient
CAS Number:377084-05-4
Formula:C₉H₇F₃O₂
MDL Number:MFCD06660214