2-Fluoro-4,6-bis(trifluoromethyl)benzamide 97%
Catalog # APOSPC302917-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:275.123 g/mol
Melting Pt:132…134 °C
Storage Temperature:Ambient
CAS Number:1017778-38-9
Formula:C₉H₄F₇NO
MDL Number:MFCD09258671