2,3-Dichloro-6-fluorophenylacetonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886497-54-7
:
Formula:C₈H₄Cl₂FN
MDL Number:MFCD04115874
:204.028 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
Frequently Bought Together