Order Entry
Czech Republic
ContactUsLinkComponent
3-Chloro-2-fluoro-4-(trifluoromethyl)phenylacetic acid 97%
3-Chloro-2-fluoro-4-(trifluoromethyl)phenylacetic acid 97%
Catalog # APOSPC302831-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-2-fluoro-4-(trifluoromethyl)phenylacetic acid 97%
Catalog # APOSPC302831-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :256.581 g/mol
Storage Temperature:Ambient
CAS Number:1431329-72-4
Formula:C₉H₅ClF₄O₂
MDL Number:MFCD23795331