
2-Ethoxy-3,5-difluorobenzoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1017779-72-4
:
Formula:C₉H₇ClF₂O₂
MDL Number:MFCD09258712
:220.6 g/mol
Storage Temperature:Ambient
UN:1760
ADR:8,III
:GHS05
Frequently Bought Together