
4-Chloro-2,6-difluorobenzoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:272104-45-7
:
Formula:C₇H₂Cl₂F₂O
MDL Number:MFCD06660235
:210.993 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together