Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Difluoro-3-methoxybenzoyl chloride
2,6-Difluoro-3-methoxybenzoyl chloride 97%
2,6-Difluoro-3-methoxybenzoyl chloride 97%
APOSPC302667-250MG
:
Apollo Scientific
:
2,6-Difluoro-3-methoxybenzoyl chloride 97%
APOSPC302667-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
206.574 g/mol
:
Ambient
:
1760
:
8,III
:
GHS05
:
886498-40-4
:
C₈H₅ClF₂O₂
:
MFCD04115903