2,6-Dichloro-3-(trifluoromethyl)benzoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:53012-82-1
:
Formula:C₈H₂Cl₃F₃O
MDL Number:MFCD11519345
:277.455 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
Frequently Bought Together