
2-Methoxy-4-(trifluoromethyl)benzyl alcohol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:286441-68-7
:
Formula:C₉H₉F₃O₂
MDL Number:MFCD04115979
:206.162 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together