
6-Chloro-2-fluoro-3-methoxybenzoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886499-63-4
:
Formula:C₈H₅Cl₂FO₂
MDL Number:MFCD04115948
ADR:8,III
:223.029 g/mol
UN:3265
:GHS05
Storage Temperature:Ambient
Frequently Bought Together