5-Chloro-2-methyl-4-(trifluoromethoxy)benzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1706447-06-4
:
Formula:C₈H₅Cl₂F₃O₃S
MDL Number:MFCD28054251
ADR:8,III
:309.09 g/mol
Storage Temperature:Ambient
UN:1760
Frequently Bought Together