Order Entry
Czech Republic
ContactUsLinkComponent
2-Methoxy-4-(trifluoromethyl)benzamide 98%
2-Methoxy-4-(trifluoromethyl)benzamide 98%
Catalog # APOSPC302389-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methoxy-4-(trifluoromethyl)benzamide 98%
Catalog # APOSPC302389-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :219.161 g/mol
Storage Temperature:Ambient
CAS Number:886500-54-5
Formula:C₉H₈F₃NO₂
MDL Number:MFCD04115978