2-Fluoro-4-(trifluoromethoxy)benzenesulphonyl chloride
Catalog # APOSPC302352-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:278.609 g/mol
Storage Temperature:Ambient
CAS Number:954388-19-3
Formula:C₇H₃ClF₄O₃S
MDL Number:MFCD28054238