4-Methoxy-3-(trifluoromethyl)cinnamic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:773131-67-2
:
Formula:C₁₁H₉F₃O₃
MDL Number:MFCD06205655
:246.183 g/mol
Melting Pt:187…189 °C
Storage Temperature:Ambient
Frequently Bought Together