
3-Fluoro-4-(trifluoromethoxy)benzoyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886499-09-8
:
Formula:C₈H₃ClF₄O₂
MDL Number:MFCD06660206
:242.555 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together