Order Entry
Czech Republic
ContactUsLinkComponent
3,5-Difluoro-2-methoxycinnamic acid 97%
3,5-Difluoro-2-methoxycinnamic acid 97%
Catalog # APOSPC302270-1G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Difluoro-2-methoxycinnamic acid 97%
Catalog # APOSPC302270-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :214.166 g/mol
Storage Temperature:Ambient
CAS Number:1092460-61-1
Formula:C₁₀H₈F₂O₃
MDL Number:MFCD09999480