Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methoxy-5-(trifluoromethoxy)benzamide
2-Methoxy-5-(trifluoromethoxy)benzamide 97%
2-Methoxy-5-(trifluoromethoxy)benzamide 97%
APOSPC302256-1G
:
Apollo Scientific
:
2-Methoxy-5-(trifluoromethoxy)benzamide 97%
APOSPC302256-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
235.16 g/mol
:
GHS07
:
Ambient
:
886499-98-5
:
C₉H₈F₃NO₃
:
MFCD04115962