
2,3-Dichloro-6-fluorobenzoyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886497-44-5
:
Formula:C₇H₂Cl₃FO
MDL Number:MFCD04115871
:227.448 g/mol
Storage Temperature:Ambient
UN:1760
ADR:8,III
:GHS05
Frequently Bought Together