Order Entry
Czech Republic
ContactUsLinkComponent
3-Fluoro-4-methoxycinnamic acid 97%
Irritant
3-Fluoro-4-methoxycinnamic acid 97%
Catalog # APOSPC302055-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Fluoro-4-methoxycinnamic acid 97%
Catalog # APOSPC302055-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:227…230 °C
 :196.175 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:713-85-9
Formula:C₁₀H₉FO₃
MDL Number:MFCD03002809