
3-(4-Chloro-2-fluorophenyl)propanoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:174603-48-6
:
Formula:C₉H₈ClFO₂
MDL Number:MFCD06656906
:202.61 g/mol
Melting Pt:83…85 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together