4-Ethoxy-2,6-difluorobenzenesulphonyl chloride
Catalog # APOSPC302024-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:256.654 g/mol
Storage Temperature:Ambient
CAS Number:1432129-18-4
Formula:C₈H₇ClF₂O₃S
MDL Number:MFCD28054226