Ethyl 2-(difluoromethyl)-6-fluoroimidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2451256-53-2
:
Formula:C₁₁H₉F₃N₂O₂
:258.197 g/mol
Melting Pt:112…114 °C
Storage Temperature:Ambient
Frequently Bought Together