6-Bromo-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2451256-52-1
:
Formula:C₉H₅BrF₂N₂O₂
:291.049 g/mol
Melting Pt:226…228 °C
Storage Temperature:Ambient
Frequently Bought Together