6-Chloro-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2451256-50-9
:
Formula:C₉H₅ClF₂N₂O₂
:246.598 g/mol
Melting Pt:218…220 °C
Storage Temperature:Ambient
Frequently Bought Together