Ethyl 2-(trifluoromethyl)imidazo[1,2-a]pyrimidine-3-carboxylate
Catalog # APOSPC301012-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:259.185 g/mol
Storage Temperature:Ambient
CAS Number:866149-79-3
Formula:C₁₀H₈F₃N₃O₂
MDL Number:MFCD05668962