1-(4-Bromo-3-fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid
Catalog # APOSPC300988-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:302.096 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₉BrFNO₃
MDL Number:MFCD21525693