2-Bromo-4-fluoro-N-methylbenzamide
Catalog # APOSPC300978-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.05 g/mol
Storage Temperature:Ambient
CAS Number:923138-87-8
Formula:C₈H₇BrFNO
MDL Number:MFCD08895950