2-(4-Bromo-3-fluorophenoxy)nicotinonitrile
Catalog # APOSPC300975-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:293.091 g/mol
Storage Temperature:Ambient
CAS Number:1516062-79-5
Formula:C₁₂H₆BrFN₂O
MDL Number:MFCD20975428