Order Entry
Czech Republic
ContactUsLinkComponent
N'-[(4-Fluoro-3-iodophenyl)methylene]imidazo[1,2-a]pyridine-7-carbohydrazide
N'-[(4-Fluoro-3-iodophenyl)methylene]imidazo[1,2-a]pyridine-7-carbohydrazide
Catalog # APOSPC300973-1G
Supplier:  Apollo Scientific
CAS Number:  
N'-[(4-Fluoro-3-iodophenyl)methylene]imidazo[1,2-a]pyridine-7-carbohydrazide
Catalog # APOSPC300973-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :408.169 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₀FIN₄O
MDL Number:MFCD31380626