Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(2-Chloro-4-fluorophenoxy)nicotinonitrile
2-(2-Chloro-4-fluorophenoxy)nicotinonitrile
2-(2-Chloro-4-fluorophenoxy)nicotinonitrile
APOSPC300972-1G
:
Apollo Scientific
:
2-(2-Chloro-4-fluorophenoxy)nicotinonitrile
APOSPC300972-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
248.64 g/mol
:
Ambient
:
1183594-45-7
:
C₁₂H₆ClFN₂O
:
MFCD13206045