N-Methyl-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
Catalog # APOSPC300954-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.19 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₀F₃N₃O
MDL Number:MFCD30481137