5-[3,5-Bis(trifluoromethyl)phenyl]-2-furonitrile
Catalog # APOSPC300953-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:305.175 g/mol
Storage Temperature:Ambient
Formula:C₁₃H₅F₆NO
MDL Number:MFCD30188174