Order Entry
Czech Republic
ContactUsLinkComponent
N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-N'-(1-tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-yl)urea
N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-N'-(1-tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-yl)urea
Catalog # APOSPC300948-1G
Supplier:  Apollo Scientific
CAS Number:  
N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-N'-(1-tetrahydro-2H-pyran-4-yl-1H-pyrazol-4-yl)urea
Catalog # APOSPC300948-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :369.342 g/mol
Storage Temperature:Ambient
Formula:C₁₆H₁₈F₃N₅O₂
MDL Number:MFCD29918613