Order Entry
Czech Republic
ContactUsLinkComponent
5-Fluoro-3-(3-hydroxybenzylidene)-1,3-dihydro-2H-indol-2-one
5-Fluoro-3-(3-hydroxybenzylidene)-1,3-dihydro-2H-indol-2-one
Catalog # APOSPC300932-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Fluoro-3-(3-hydroxybenzylidene)-1,3-dihydro-2H-indol-2-one
Catalog # APOSPC300932-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :255.244 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₀FNO₂
MDL Number:MFCD30481171