Order Entry
Czech Republic
ContactUsLinkComponent
4-[2-Methyl-3-(trifluoromethyl)phenyl]-2,4-dihydro-3H-1,2,4-triazol-3-one
4-[2-Methyl-3-(trifluoromethyl)phenyl]-2,4-dihydro-3H-1,2,4-triazol-3-one
Catalog # APOSPC300920-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[2-Methyl-3-(trifluoromethyl)phenyl]-2,4-dihydro-3H-1,2,4-triazol-3-one
Catalog # APOSPC300920-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :243.185 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₈F₃N₃O
MDL Number:MFCD30481160