N-(2,4-Dibromo-6-fluorophenyl)-N'-methylthiourea
Catalog # APOSPC300904-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:342.026 g/mol
Storage Temperature:Ambient
Formula:C₈H₇Br₂FN₂S
MDL Number:MFCD07115988