N-[4-Bromo-2-(trifluoromethoxy)phenyl]-N'-methylthiourea
Catalog # APOSPC300903-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:329.137 g/mol
Storage Temperature:Ambient
CAS Number:1988424-15-2
Formula:C₉H₈BrF₃N₂OS
MDL Number:MFCD30188166