Order Entry
Czech Republic
ContactUsLinkComponent
4-[8-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]benzonitrile
4-[8-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]benzonitrile
Catalog # APOSPC300863-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[8-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]benzonitrile
Catalog # APOSPC300863-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :287.239 g/mol
Storage Temperature:Ambient
CAS Number:1980053-08-4
Formula:C₁₅H₈F₃N₃
MDL Number:MFCD28992255