Order Entry
Czech Republic
ContactUsLinkComponent
4-[3-(Difluoromethoxy)phenyl]-2,4-dihydro-3H-1,2,4-triazol-3-one
4-[3-(Difluoromethoxy)phenyl]-2,4-dihydro-3H-1,2,4-triazol-3-one
Katalogové číslo APOSPC300826-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[3-(Difluoromethoxy)phenyl]-2,4-dihydro-3H-1,2,4-triazol-3-one
Katalogové číslo APOSPC300826-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :227.168 g/mol
Storage Temperature:Ambient
CAS Number:1858255-65-8
Formula:C₉H₇F₂N₃O₂
MDL Number:MFCD28148247