
N-[2-(2-Fluorophenoxy)benzyl]-N-methylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1095045-47-8
:
Formula:C₁₄H₁₄FNO
MDL Number:MFCD11631315
:231.265 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together