
2,3,5-Trifluoro-4-(4-methylpiperazin-1-yl)benzoic acid
Katalogové číslo APOSPC300806-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:274.239 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:812643-49-5
Formula:C₁₂H₁₃F₃N₂O₂
MDL Number:MFCD28103355