Order Entry
Czech Republic
ContactUsLinkComponent
4-Bromo-2-(2,2-diethoxyethoxy)-1-fluorobenzene
4-Bromo-2-(2,2-diethoxyethoxy)-1-fluorobenzene
Catalog # APOSPC300794-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-2-(2,2-diethoxyethoxy)-1-fluorobenzene
Catalog # APOSPC300794-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :307.156 g/mol
Storage Temperature:Ambient
CAS Number:1777895-41-6
Formula:C₁₂H₁₆BrFO₃
MDL Number:MFCD26939171