Methyl 2,5-difluoro-4-(4-methylpiperazin-1-yl)benzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1858251-84-9
:
Formula:C₁₃H₁₆F₂N₂O₂
MDL Number:MFCD28103349
:270.275 g/mol
Storage Temperature:Ambient
Frequently Bought Together